missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Chelating Sepharose FF

Hitrap chelating sepharose FF

Brand:  Cytiva 90100480

Product Code. 15519254

  • £1046.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • HiTrap Chelating Sepharose FF
  • Storage: 4 to 30°C, 20% Ethanol
  • Supplied in packs of 5 x 5mL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

4°C to 30°C. 20% Ethanol
7 mm
1 mL
5 x 1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

SDS
Documents

Documents

Certificates
Special Offers

Special Offers