missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ CM Sepharose Fast Flow IEX Columns
Weak cation exchanger for small-scale protein purifications
£136.00 - £299.28
Specifications
Description | HiTrap CM FF |
---|---|
For Use With (Application) | For method scouting, group separations, sample concentration and sample clean-up of charged biomolecules |
Content And Storage | 4°C to 30°C in 20% Ethanol |
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
10420695
|
Cytiva
17-5056-01 |
1 mL | 5 x 1 mL |
£136.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11525025
|
Cytiva
17-5155-01 |
5 mL | 5 x 5 mL |
£299.28
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Convenient for rapid separations either alone or connected in series
- Designed for use with syringe, peristaltic pump or chromatographic system
- High flowrates and good scale-up potential
Specifications
HiTrap CM FF | |
For method scouting, group separations, sample concentration and sample clean-up of charged biomolecules | |
4°C to 30°C in 20% Ethanol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- hitrap CM FF
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title