Learn More
Cytiva HiTrap™ Desalting Prepacked Columns
For rapid convenient desalting and buffer exchange. HiTrap™ Desalting Prepacked Columns are prepacked, ready to use column for group separation between high and low molecular weight substances.
Brand: Cytiva 17-1408-01
228.00 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : GE
Additional Details : Weight : 0.13500kg
Description
- Prepacked with Sephadex™ G-25 Superfine media
- Column material: polypropylene
- Bed volume: 5mL
- Bed dimensions: 16 x 25mm
- Sample volume: 1.5mL
- Flow velocity: <15mL/min.
- Maximum pressure: 3 bar
Media
- Matrix: cross-linked dextran
- Partical size: >20μm
- pH stability working range: 2 to 13
Specifications
HiTrap Desalting are ready-to-use 5 mL columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange. | |
>20 μm (dry), 15 to 88 μm (wet) | |
For rapid convenient desalting and buffer exchange | |
4°C to 30°C in 20% Ethanol | |
No |
HiTrap Desalting, 5 x 5 mL | |
0.62 in. | |
5 mL | |
16 mm |