missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Desalting Prepacked Columns

Product Code. p-3676561 Shop All Cytiva Products
Click to view available options
Product Type:
HiTrap Desalting, 5 x 5 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 11525015

Brand: Cytiva 17140801

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For rapid convenient desalting and buffer exchange. HiTrap™ Desalting Prepacked Columns are prepacked, ready to use column for group separation between high and low molecular weight substances.

  • Prepacked with Sephadex™ G-25 Superfine media
  • Column material: polypropylene
  • Bed volume: 5mL
  • Bed dimensions: 16 x 25mm
  • Sample volume: 1.5mL
  • Flow velocity: <15mL/min.
  • Maximum pressure: 3 bar

Media
  • Matrix: cross-linked dextran
  • Partical size: >20μm
  • pH stability working range: 2 to 13

TRUSTED_SUSTAINABILITY

Specifications

Description HiTrap Desalting are ready-to-use 5 mL columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange.
Product Type HiTrap Desalting, 5 x 5 mL
Particle Size >20 μm (dry), 15 to 88 μm (wet)
Diameter (English) Inner Column 0.62 in.
For Use With (Application) For rapid convenient desalting and buffer exchange
Volume (Metric) Bed 5 mL
Content And Storage 4°C to 30°C in 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Disposable Reusable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.