Learn More
Cytiva HiTrap™ HIC Sepharose™ Selection Kit
Use for laboratory and intermediate process-scale separations and final-step purifications. Cytiva Life Sciences™ HiTrap™ HIC Sepharose™ Selection Kit takes full advantage of hydrophobic interaction chromatography providing highest resolution.
Brand: Cytiva 28-4110-07
Code : A0
Additional Details : Weight : 0.11700kg
Description
- Includes seven 1mL HIC columns, each packed with a different medium for small-scale media screening and selection
- Flexible design for use with syringes, single laboratory pump or chromatography system
- High flowrates and high capacities
Media in Kit Columns:
Phenyl Sepharose High Performance
- For laboratory and intermediate process-scale separations and final-step purifications needing a high resolution
- For intermediate and final purification steps
- Relatively high ligand density enables high dynamic binding capacity for proteins, even at low salt concentrations
- For initial and intermediate step purifications requiring a matrix with low to medium hydrophobicity
- Often works well even in relatively low salt concentrations
- Least hydrophobic option in the Fast Flow series
- Suited for binding and elution of relatively strong hydrophobic proteins
- Different hydrophobic character from the phenyl and butyl ligands
Specifications
1 mL | |
7 mm | |
< 4 mL/min | |
Seven HiTrap 1 mL column containing the following media: Phenyl Sepharose high performance, Butyl Sepharose high performance, Phenyl Sepharose 6 fast flow (low sub), Phenyl Sepharose 6 fast flow (high sub), Butyl-S Sepharose 6 fast flow, Butyl Sepharose 4 | |
5 bar [0.5 MPa] (70 psi) |
0.275 in. | |
HiTrap HIC Selection Kit | |
7 x 1 mL | |
Polypropylene | |
4°C to 30°C, 20% Ethanol |
Safety and Handling
- HiTrap HIC Selection Kit'
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol, ethanol, ethanol, ethanol, ethanol, ethanol, ethanol