Learn More
Cytiva HiTrap™ IgY Purification Column
Developed for fast, easy purification of IgY from chicken egg yolk
Brand: Cytiva 17-5111-01
Includes: Complete instructions and a variety of connectors
Description
- Prepacked with a thiophilic adsorption medium, 2-mercaptopyridine coupled to Sepharose™ High Performance medium
- Each column binds one-quarter egg yolk (100mg); columns can be connected in series for greater capacity
- Excellent recovery (approx. 80%) and purity (usually >70%)
- Designed for use with a syringe, peristaltic pump, or chromatography system

Specifications
1.6 cm | |
<20 mL/min. | |
Antibody affinity chromatography | |
20% Ethanol | |
5 mL |
0.62 in. | |
5 mL | |
1 mL | |
4°C to 30°C, 20% Ethanol |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- HiTrap IgY Purification 5 ml, 1 x 5 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.