missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap KappaSelect Media
Shop All Cytiva Products
Click to view available options
Bed Volume (Metric):
1 mL
5 mL
Unit Size:
Each
Pack of 5
Specifications
Specifications
| Quantity | 5 x 1 mL |
| Stability | Stable in all commonly used aqueous buffers |
| Storage Requirements | 4 to 8 deg.C, 20% Ethanol |
| Binding Capacity | 15 mg Fab/mL of medium2) |
| Product Type | HiTrap KappaSelect Media |
| Particle Size | 75 μm |
| Material | Polypropylene |
| Max. Pressure | 300 kPa |
| Matrix | Highly Cross-linked Agarose |
| pH Range | 3 to 10 |
| Show More |
Safety and Handling
Product Identifier
- HiTrap 1 x 5 ml, KappaSelect
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction