missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Phenyl FF (high sub) Columns

Prepacked columns for convenient and fast separations

Brand:  Cytiva 17-1355-01


 View more versions of this product

Product Code. 10094194

  • £188.00 / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Flexible design for use with a syringe, laboratory pump or chromatography system
  • Available in 1mL and 5mL sizes
  • High flowrates and high capacities
  • Easy scale-up achieved by connecting columns in series

  • Maximum flowrates: 4mL/min. for 1mL columns and 20mL/min. for 5mL columns

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
7 mm
< 1 mL/min
5 x 1 mL
25 mm
Polypropylene
5 bar [0.5 MPa] (70 psi)
3 to 13
4°C to 30°C, 20% Ethanol
0.275 in.
HiTrap Phenyl FF (High Sub)
For Screening of Selectivity, Binding and Elution Conditions, as Well as Small Scale Purifications
0.984 in.
Phenyl
Phenyl Sepharose 6 Fast Flow (high sub)
45 to 165 μm
6% Cross-linked Agarose
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Phenyl FF (high sub) 1 ml, 5 x 1 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers