missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Protein G HP Prepacked Columns

Product Code. p-3667288 Shop All Cytiva Products
Click to view available options
Binding Capacity:
125 mg Human IgG/Column
25 mg Human IgG/Column
Diameter (English) Inner Column:
0.27 in.
0.62 in.
Flow Rate:
>20 mL/min.
>4 mL/min.
<4 mL/min.
Volume (Metric) Bed:
1 mL
5 mL
Unit Size:
1mL
Each
Pack of 2
Pack of 5
This item is not returnable. View return policy

Product Code. 10185894

Brand: Cytiva 17040401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for fast, easy separations of IgG from different species and subclasses. Cytiva Life Sciences™ HiTrap™ Protein G HP Prepacked Columns are prepacked with Protein G Sepharose™.

  • Prepacked with Protein G Sepharose High Performance
  • Binding capacity: 25mg human IgG/column
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • For use with syringe, peristaltic pump, or chromatography system
  • 1 and 5mL columns

Media
  • Ligand: Recombinant protein G lacking albumin-binding region
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: 3 to 9

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Pre-packed Columns
Binding Capacity 25 mg Human IgG/Column
Diameter (Metric) Inner Column 7 mm
Diameter (English) Inner Column 0.27 in.
Flow Rate <4 mL/min.
Volume (Metric) Bed 1 mL
For Use With (Application) Designed for use with a syringe, peristaltic pump, or chromatography system
Description HiTrap Protein G HP are prepacked columns for general-purpose preparative purification of monoclonal and polyclonal antibodies from most species including rat.
Storage Buffer 20% Ethanol
Content And Storage 4°C to 30°C
Quantity 5 Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.