Learn More
Cytiva HiTrap™ Q Sepharose Fast Flow IEX Columns
Use for small-scale protein purifications. Cytiva Life Sciences™ HiTrap™ Q Sepharose Fast Flow IEX Columns are strong anion exchangers for rapid separations either alone or connected in series.
Brand:  Cytiva 17-5156-01
 View more versions of this product
Product Code. 11535025
  • £299.00 / 25mL
Edge
Explore more special offers
Description
Description
  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HiTrap Q FF
<20 mL/min.
4°C to 30°C in 20% Ethanol
5 mL
0.6 in.
HiTrap Q FF is a strong anion exchange chromatography column for small-scale protein purification.
16 mm