missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Q Sepharose XL IEX Columns
Optimized for fast, convenient small-scale protein capture
Brand: Cytiva 17-5159-01
This item is not returnable.
View return policy
Description
- Designed for use wtih a syringe, peristaltic pump or chromatography system
- For group separations, concentrations and sample cleanup
- Convenient and affordable for fast, easy separations either alone or connected in series
- High binding capacities with high flowrates
- Media consists of long chains of dextran coupled to a robust, 6% highly cross-linked agarose matrix
- Dextran chains increase the exposure of charged groups for high loading capacities
Specifications
HiTrap Q XL | |
<20 mL/min. | |
4°C to 30°C in 20% Ethanol | |
5 mL |
0.6 in. | |
HiTrap Q XL are strong anion exchangers prepacked with Q Sepharose XL media, optimized for fast, convenient small-scale protein capture using ion exchange (IEX) chromatography. | |
16 mm | |
5 x 5 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Q XL 5 ml, 5 x 5 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction