missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ SP Sepharose XL IEX Columns
Optimized for fast, convenient small-scale protein capture
£134.00 - £295.00
Specifications
Description | HiTrap SP XL |
---|---|
For Use With (Application) | HiTrap SP XL are strong cation exchangers prepacked with SP Sepharose XL media, optimized for fast, convenient small-scale protein capture using ion exchange (IEX) chromatography. |
Content And Storage | 4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol |
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
10480705
|
Cytiva
17-5160-01 |
1 mL | 1 x 5 mL |
£134.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11565025
|
Cytiva
17-5161-01 |
5 mL | 5 x 5 mL |
£295.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Designed for use wtih a syringe, peristaltic pump or chromatography system
- For group separations, concentrations and sample cleanup
- Convenient and affordable for fast, easy separations either alone or connected in series
- High binding capacities with high flowrates
- Media consists of long chains of dextran coupled to a robust, 6% highly cross-linked agarose matrix
- Dextran chains increase the exposure of charged groups for high loading capacities
Specifications
HiTrap SP XL | |
HiTrap SP XL are strong cation exchangers prepacked with SP Sepharose XL media, optimized for fast, convenient small-scale protein capture using ion exchange (IEX) chromatography. | |
4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap SP XL 1 ml, 5 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol