missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ HIV-1 Env gp160 Microarray

HIV-1 Env gp160 Microarray with 856 protein specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.014.001

Additional Details : Weight : 0.02200kg

Product Code. 16367758

  • £482.30 / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket

Description

Description

  • Microarray Content: Envelope glycoprotein (Env) gp160 of HIV type 1 cleaved into 2 chains: Surface protein gp120 and Transmembrane protein gp41
  • Peptide Length/Overlap: 15 aa/ 14 aa
  • Number of Peptides: 856 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEPperSlide™ PEGMA/PMMA
Antibody validation, epitope mapping, serum screening, biomarker discovery
Microscopic Glass
Individual barcode for each microarray
1 Slide
1 mm
Transparent
75.4 x 25 mm
High-density Peptide Microarray
Non-sterile
3 array copies per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers