missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14336-1-AP-150UL
This item is not returnable.
View return policy
Description
CD98 (4F2) is a type II transmembrane glycoprotein which serves as the heavy chain of the heterodimeric amino acid transporters (HATs). CD98, linked to various light chains by disulfide bond, is responsible for cell surface expression and basolateral localization of this transporter complex in polarized epithelial cells and also interacts with beta1 integrins and increases their affinity for ligand. Besides its roles in amino acid transport, CD98 is thus involved in cell fusion and activation. It is implicated in regulation of cellular differentiation, growth and apoptosis.Specifications
HLX | |
Polyclonal | |
Unconjugated | |
HLX | |
H2.0 like homeobox, H2.0 like homeobox protein, HB24, HLX, HLX1, Homeobox protein HB24, Homeobox protein HLX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15284, 3142, 364069 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A0JPN1, Q14774, Q61670 | |
HLX | |
HLX Fusion Protein Ag5627 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |