missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ HM 430, HM 450 and 440 E Sliding Microtome Accessories
Maintain Epredia™ HM 430 and HM 450 sliding microtomes with Epredia™ HM 430, HM 450 and 440 E sliding microtomes accessories.
Brand: Epredia™ 715480
586.50 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
Standard buffer tray
Cooling element for buffer solution
Large-field magnifier
StemiScope™ with optics and mount
Swan-neck light guide
Specifications
Standard Specimen Clamp with Waste Tray, 45 x 80 mm | |
Sliding Microtome Accessories | |
Epredia HM 430 and HM 450 Sliding Microtomes |