Learn More
Epredia™ HM 525 NX Cryostat Blade Holders
Use Epredia™ HM 525 NX blade holders with HM 525 NX and other Epredia™ cryostats.
£885.00 - £2265.00
Produktcode | Marke | Description | For Use With (Equipment) | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Description | For Use With (Equipment) | Preis | Menge & Verfügbarkeit | ||||
12053079
|
Epredia™
566790 |
Standard Knife Carrier SM and SMV | Microm HM 560 Cryostat | N/A | |||||
12013109
|
Epredia™
705460 |
Standard Knife Holder | Epredia HM 525 NX Cryostat and older Microm HM 550 |
£1480.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12023109
|
Epredia™
705470 |
Disposable Blade Carrier EC | Epredia HM 525 NX Cryostat and older Microm HM 550 |
£1560.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12053109
|
Epredia™
705580 |
Disposable Blade Carrier EV | Epredia HM 525 NX Cryostat and older Microm HM 550 | N/A | |||||
12063109
|
Epredia™
705630 |
Disposable Blade Carrier EC70 | Epredia HM 525 NX Cryostat and older Microm HM 550 |
£2265.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
Standard Knife Carrier:
- Allows for insertion of standard knives, with clearance angle adjustment
- Can be moved horizontally on precision dovetail guide with quick clamp system
- Includes anti-roll plate
Disposable Blade Carrier EC:
- Allows for insertion of high- and low-profile blades, with clearance angle adjustment
- Can be moved on precision dovetail guide with quick clamp plate
- Includes anti-roll plate
Disposable Blade Carrier EV:
- Use with vacutome system
- Allows for insertion of high- and low-profile disposable blades
- Includes a vacuum anti-roll hood for stretching and disposing of section waste
- Features a slot that is opened and closed by a vacuum-controlled flap
Magnetic Disposable Blade Carrier MV:
- Use with vacutome system
- Allows for insertion of disposable blades
- Includes a vacuum anti-roll hood for stretching and disposing of section waste
- Features a slot that is opened and closed by a vacuum-controlled flap
Spezifikation
Cryostat Blade Holder |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts