Learn More
HO-1/HMOX1 Mouse anti-Human, CoraLite 488, Clone: 2D10A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66743-100UL
This item is not returnable.
View return policy
Description
The HO-1/HMOX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HO-1/HMOX1. This antibody recognizes human, mouse, rat, pig antigen. The HO-1/HMOX1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
HO-1/HMOX1 | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
3162 | |
IgG2a |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
2D10A5 | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |