missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Hoechst 34580
Hoechst 34580
Brand: Molecular Probes™ H21486
This item is not returnable.
View return policy
Description
This bisbenzimide dye, Hoechst 34580, is related to the important minor-groove binding, dsDNA-selective dyes Hoechst 33258 and Hoechst 33342. Hoechst 34580 has somewhat longer-wavelength spectra than the other Hoechst dyes when bound to nucleic acids [Howard Shapiro, personal communication].Specifications
Contains 1 vial of Hoechst 34580 (5 mg). Store in freezer -25°C and protect from light. |
|
Hoechst 34580 | |
Fluorescence Microscope | |
Fluorescence | |
Room Temperature | |
Nucleic Acid Stain |
392⁄440 | |
Tube(s) | |
5 mg | |
Solid | |
water, DMF or DMSO (Dimethylsulfoxide) | |
Nucleic Acids, Nucleus |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction