missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Air-Cooled Vertical Electrophoresis Unit

Compact, rugged, economical unit ideal for educational labs with multiple users

Brand:  Hoefer™ SE400

Product Code. 12865836

  • £982.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Lower buffer chamber and casting stand on leveling base, upper buffer chamber, lid with cables, two 18 x 16cm glass plates, two 16cm clamp assemblies, two cams, gel seal.
This item is not returnable. View return policy

Description

Description

This air-cooled, low-cost unit is ideal for student labs and for labs requiring many units. It also makes affordable the need to run gels under differing conditions when temperature control is not crucial.

  • Sturdier vertical slab gel unit
  • Runs one 18 x 16 cm gel at a time or two at a time with accessory Club Sandwich Kit
  • Trouble-free to set up and run; safe to operate
  • Precision-molded parts fit together easily
  • Assembling gel sandwiches takes two minutes
  • Sandwiches seal leak-free without the use of tape or grease in the built-in casting stand
  • Specially designed safety lid prevents accidental contact with buffer
  • Rugged injection-molded construction means the unit will last a long time
  • Plates, clamps, spacers, and combs are interchangeable with the SE600 accessories
  • Club sandwich feature saves space and money; with special divider plate, it runs two 18 x 16 cm gels at a time
  • Combs and spacers sold separately
  • Plate size: 18 x 16 cm
  • Spacers: 0.75, 1.0 and 1.5 mm
  • Maximum power settings: 500V, 60mA and 20W
  • Dimensions: 10 x 9.4 x 5 in. (28 x 24 x 13 cm)
Specifications

Specifications

EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Lower buffer chamber/casting stand on leveling base with high voltage leads, upper buffer chamber, safety lid with electrodes, glass plates, clamp assemblies, cams, slotted gasket for upper buffer chamber, bubble level, laminated gasket for casting stand, spacer mate alignment template, wonder wedge-plate separation tool, gel seal
45°C
3.66 oz.
9.44 in.
24 cm
11.02 in.
28 cm
15 cm
20 w
Up to 80%
113°F
15°C to 30°C
700 mL
7.08 in.
18 cm
6.29 in.
5.9 in.
500 V
60 mA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers