missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Buffer Dam for SE 600 Series Electrophoresis Unit
Buffer Dam for SE 600 Series Electrophoresis Unit
Brand: Hoefer™ SE6032
Code : A0
Additional Details : Weight : 0.80000kg
Description
- For use with SE 600 Series Electrophoresis Unit
Specifications
SE 600 Series Electrophoresis Unit | |
Buffer Dam |