Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Brand:  Hoefer™ SE211A-R-1.5
 View more versions of this product
Product Code. 12004657
  • £47.01 / Each
Estimated Shipment: 11-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
  • Lightweight
  • Virtually unbreakable
Specifications
Specifications
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
PTFE
0.05 in.
120μL/ mm
6.8 cm
1.3 cm
Comb
1 Prep, 1 Reference
0.15 cm
2.6 in.
0.51 in.