missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Brand: Hoefer™ SE511-20-1,5
This item is not returnable.
View return policy
Description
- Lightweight
- Virtually unbreakable
- Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
- Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth

Specifications
SE 600, SE 400 Series Vertical Electrophoresis System | |
PTFE | |
0.05 in. | |
6.2μL/ mm | |
0.98 in. |
Standard Comb | |
20 | |
0.15 cm | |
0.41 cm | |
2.5 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units > Comb; 20 wells; 1.5mm thick
Spot an opportunity for improvement?Share a Content Correction