Learn More
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Brand: Hoefer™ SE511-20-.75
Description
- Lightweight
- Virtually unbreakable
- Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
- Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth
Specifications
SE 600, SE 400 Series Vertical Electrophoresis System | |
PTFE | |
0.02 in. | |
3.1μL/ mm | |
0.98 in. |
Standard Comb | |
20 | |
0.07 cm | |
0.41 cm | |
2.5 cm |