missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Brand:  Hoefer™ SE511-20-1.5

 View more versions of this product

Product Code. 12069807

  • £96.55 / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Lightweight
  • Virtually unbreakable
  • Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
  • Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth
Specifications

Specifications

SE 600, SE 400 Series Vertical Electrophoresis System
PTFE
0.05 in.
6.2μL/ mm
0.98 in.
Standard Comb
20
0.15 cm
0.41 cm
2.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units > Comb; 20 wells; 1.5mm thick

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.