Learn More
Hoefer™ Easy Breeze™ Gel Dryer Accessories: Film
For use with the SE 1200 Easy Breeze Gel Drying System
Brand:  Hoefer™ SE1202
Product Code. 10418254
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Gel Drying Film
  • For photography or fluorography, clear cellophane ensures a photograph or exposure without distortion
  • For autoradiography, use of a Mylar™ sheet in place of one cellophane sheet allows the Mylar to be easily removed after drying for maximum beta emission
Specifications
Specifications
Standard Frame of the SE 1200 Easy Breeze™ Air Gel Drying System
Porous Cellophane Sheet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hoefer™ Easy Breeze™ Gel Dryer Accessories: Film > Porous cellophane sheets