Learn More
Hoefer™ Gel Casters for Mini Vertical Electrophoresis Systems
Easy-to-use casters
Brand:  Hoefer™ SE235
 View more versions of this product
Product Code. 10772677
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
SE245 Dual-Gel Caster
  • For use with SE250, SE260, or MiniVE Vertical Electrophoresis Systems
  • SE235 4-Gel Caster, SE275 4-Gel Caster, and SE215 10-Gel Caster
    • Suitable for single concentration and gradient gels
    • SE235 4-Gel Caster is for use with SE260 or MiniVE Electrophoresis Systems
    • SE 275 4-Gel Caster and SE215 10-Gel Caster are for use with SE250 Electrophoresis System
    TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    SE 260, MiniVE™ Electrophoresis System
    Gel Caster
    Caster body with gasket, face plate with inlet port, 10 x 10.5cm rectangular Glass plates, 10 x 10.5cm notched Alumina plates, spring clamps, Wax Paper, space-saver plate, Polycarbonate filler sheets, black Vinyl cap, filler plug set
    1 to 4