Learn More
Hoefer™ Gel Casters for SE 600 and SE 400 Series Vertical Electrophroesis Systems
Cast multiple gels
Brand:  Hoefer™ SE6015
Product Code. 12044877
  • £325.00 / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easy-to-handle
  • Leak-free
  • Suitable for one or two gels
Specifications
Specifications
SE 600, SE 400 Series Vertical Electrophoresis System
Caster stand with 2 cradles and leveling feet, laminated gaskets
7.08 in.
Dual Gel Caster
2
18 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products