missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ HE 33 Mini Submarine Electrophoresis Unit
Designed for very fast separations of DNA restriction fragments in agarose gels
£368.28 - £499.00
Specifications
Certifications/Compliance | EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE |
---|---|
Type | Mini Horizontal Agarose Electrophoresis Unit |
Humidity Range | Up to 80% |
Sample Size | Up to 32 |
Temperature (English) Maximum | 122°F |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10646253
|
Hoefer™
HE33B |
Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level |
£368.28
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
11589533
|
Hoefer™
HE33-8-1.5 |
Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level, 1.5mm thick, 8-well comb, comb back |
£372.20
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
10754807
|
Hoefer™
HE33-8-1.5 |
Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level, 1.5mm thick, 8-well comb, comb back |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10075904
|
Hoefer™
HE33B |
Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Designed for very fast separations of DNA restriction fragments in agarose gels
- The clear molded base acts as a heat-sink—simply fill the base with coolant and seal
- Cooling the unit prior to a run allows higher voltages to be applied without generating band distortions or damaging the gel and unit
- No taping required for casting
- Electrical safety locks in lid
Specifications
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE | |
Up to 80% | |
122°F | |
8.4 oz. | |
9.4 x 5.1 x 2.8 in. (24 x 13 x 7 cm) | |
3.9 in. | |
10 cm | |
2.7 in. | |
7 cm | |
7 cm | |
15 w |
Mini Horizontal Agarose Electrophoresis Unit | |
Up to 32 | |
50°C | |
250 mL | |
9.4 in. | |
24 cm | |
5.1 in. | |
13 cm | |
2.8 in. | |
500 V | |
500 mA |