missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ HE 99X Max Submarine Electrophoresis Unit
Offers no-mess gel casting without tape or dams
£470.15 - £611.19
Specifications
Certifications/Compliance | EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE |
---|---|
Type | Horizontal Agarose Electrophoresis Unit |
Humidity Range | Up to 80% |
Sample Size | 60 |
Temperature (English) Maximum | 113°F |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11519573
|
Hoefer™
HE99X |
Buffer chamber assembly, safety lid with high voltage Leads, Foam gaskets, bubble level |
£470.15
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
12004817
|
Hoefer™
HE99X-15-1.5 |
Buffer chamber assembly, safety lid with high voltage Leads, 15 x 20cm running tray, 15 x 20cm casting tray, Foam gaskets, bubble level, 1.5mm thick, 15-well comb, Comb Back |
£611.19
Each |
Estimated Shipment: 13-11-2024
Log in to see stock. |
|||||
Description
- Unit accommodates a variety of gel lengths for maximum resolution of DNA fragments
- Agarose bridges are also accommodated, so gels can be run in nonsubmarine fashion
- Rugged design provides reliability and safety
Specifications
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE | |
Up to 80% | |
113°F | |
40.5 oz. | |
14.1 x 7.1 x 5.5 in. (36 x 18.2 x 14 cm) | |
5.9, 3.9 or 7.8 in. | |
15, 10 or 20 cm | |
5.9 in. | |
15 cm | |
14 cm | |
20 w |
Horizontal Agarose Electrophoresis Unit | |
60 | |
45°C | |
1200 mL | |
14.1 in. | |
36 cm | |
7.1 in. | |
18.2 cm | |
5.5 in. | |
200 V | |
100 mA |