Learn More
Hoefer™ High-Voltage Lead Set
Accessory for Electrophoresis Unit
Brand:  Hoefer™ SE6056-HV
Additional Details : Weight : 0.40000kg
Product Code. 15508991
  • £146.53 / 1 set
Estimated Shipment: 13-08-2024
to see stock.
Add to basket
Description
Description
  • High-voltage Lead Set (1 red and 1 black)
  • Female end is for use with Hoefer safety lids only
  • Male end is sheathed and may require an adaptor to fit power source from another manufacturer
Specifications
Specifications
High Voltage Lead Set
One red, one black