missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Mighty Small™ II Mini Vertical Electrophoresis Systems
Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids
£666.04 - £713.05
Product Code | Brand | Height (Metric) | Length (Metric) Gel | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Length (Metric) Gel | Includes | Price | Quantity & Availability | |||
11925358
|
Hoefer™
SE250 |
16 cm | 8 cm | Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, two 10 x 8cm notched Alumina plates, four spring clamps, gel seal, well locating decal and spacer mate |
£666.04
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
|||
11935358
|
Hoefer™
SE260B |
18 cm | 8/10.5 cm | Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, four spring clamps, gel seal, well locating decal and spacer mate |
£666.04
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||
10137294
|
Hoefer™
SE260-10A-.75 |
18 cm | 8/10.5 cm | Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers (0.07 cm), well locating decals, gel seal |
£705.22
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||
15182345
|
Hoefer™
SE260-10A-1.5 |
18 cm | 8/10.5 cm | Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers (0.15 cm), well locating decals, gel seal |
£713.05
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||
11519683
|
Hoefer™
SE250-10A-75 |
16 cm | 8 cm | Mini Vertical Electrophoresis Unit, complete with 0.75mm Accessories |
£697.38
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||
13409436
|
Hoefer™
SE250-10A-1.0 |
18 cm | 8 cm | Mini Vertical Electrophoresis Unit, complete with 1mm Accessories |
£697.38
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||
15192345
|
Hoefer™
SE250-10A-1.5 |
18 cm | 8 cm | Mini Vertical Electrophoresis Unit, complete with 1.5mm Accessories |
£697.38
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||
Description
Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids
- Run two gels at once
- Produce high-resolution separations in as little as 45 minutes with no band “smiling”
- Use alumina plates (included with specified models) for efficient cooling
- Buffer volume: 300 mL
Applications:
Ideal for uniform and gradient acrylamide, agarose, IEF, field inversion agarose and 2-D electrophoresis
Certifications
CE Marked, CSA C22.2 1010.1, EN61010-1, UL3101-1.