missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Mini-Peristaltic Pump
For use with horizontal units with buffer recirculation ports
Brand: Hoefer™ PP24
This item is not returnable.
View return policy
Description
The Mini-Peristaltic Pump can be used with either a single tube or two tubes simultaneously. Two of the Pump Head Tubing Pieces are included with the pump. Additional Pump Head Tubing Pieces may be purchased separately.
- Two front panel controls provide flow rates from approximately 0.8 to 24.5mL/min.
- Control knob provides variable adjustment from 0 to 100% of the selected flow rate range
- Second control is a two position toggle switch marked x1, x2 which selects low or high flow rates
- Four roller pump head is on top of the stout metal box
- Back of the pump head rotates into an“open”position and either one or two tubes can be dropped into slots
- Loaded section rotates back against spring loaded jaws and locks into place
- Tubing is automatically in proper wiping contact with the pump head rollers
- Each pump is provided with a 12.5mm (0.5 in.) rod clamp on the back so that multiple pumps can be mounted vertically on a lattice rod
Specifications
EN61010-1, CE | |
Mini Peristaltic Pump | |
41.3 in. | |
44.88 in. | |
189 cm | |
10 w |
Horizontal Unit with Buffer Recirculation Port | |
41.3 x 44.88 in. (105 x 114 cm) | |
105 cm | |
114 cm | |
115/230 V | |
800 mA |