Learn More
Hoefer™ miniVE Electrophoresis Unit Accessories and Parts
Part and accessories for miniVE Electrophoresis Unit
Brand:  Hoefer™ SE309
Code : NEW
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11975378
  • £182.57 / Each
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
  • Accesories include High Voltage Leads (2 pcs), Banana Plug Kit (2 plugs, 2 nuts, 4 washers), Foam Gasket, Clamp Screws (5 pcs), Upper Buffer Chamber Assembly, Clamp Assembly, Sealing Plate Assembly, Sealing Gasket and Lower Buffer Chamber (tank)
Specifications
Specifications
MiniVE Vertical Electrophoresis System
Upper Buffer Chamber Assembly
Gasket and banana plug kit