Learn More
Hoefer™ miniVE Integrated Vertical Electrophoresis
Perform both electrophoresis and electro transfer in one sturdy, compact unit.
Brand:  Hoefer™ SE300
Additional Details : Weight : 2.40000kg
Product Code. 15588971
  • £720.89 / 1 set
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
  • Basic
Specifications
Specifications
Protein electrophoresis, Electro transfer (blotting)
Lower Buffer Chamber (tank); Gel Modules with Gaskets, 2 assembliesl Safety Lid with High Voltage Leads; Well-Locating Decals, 2 pcs; Wonder Wedge Plate Separation Tool Gel Seal
32 x 37 x 14 cm
Up to 80%
Basic