missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Replacement Parts for HE 33 Electrophoresis Unit: Buffer Chamber Assembly
For use with HE 33 Electrophoresis Units
Brand: Hoefer™ HE30
Code : NEW
Additional Details : Weight : 0.23000kg
Description
- Buffer Chamber Assembly
Specifications
HE33 Mini Submarine Unit | |
Buffer Chamber Assembly | |
Buffer chamber (base) with feet, Platinum electrode wires and banana plug |