missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ SE 900 Large Format Vertical Electrophoresis System
Ideal for use with 24cm IPG strips in the second dimension of 2-D electrophoresis
£7415.00
Specifications
Certifications/Compliance | EN61010-1, UL61010-1, CSA C22.2, CE |
---|---|
Humidity Range | Up to 80% |
Temperature (English) Maximum | 113°F |
Temperature (Metric) Maximum | 45°C |
Temperature (Metric) Storage | 15°C to 30°C |
Includes: Buffer chamber with pumps, safety lid with leads, gel rack, 10 ft. tubing, two pair recirculation bath connectors, drain connector, seven tubing clamps, gel seal, wonder wedge plate separation tool
Mfr. No. SE900-1.0 also includes: Multicaster with ported face plate, six red spring clamps, two thumb screws, ten divider sheets, filler plug, cassette blank, gel seal, six glass hinge casettes
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11995368
|
Hoefer™
SE900-1.0 |
Separation tank, tubing kit, lid with high voltage Leads, multiple gel caster, internal PAGE rack, hinged Glass cassettes |
£7415.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Runs 1 to 6 gels
- Uses hinged cassettes for convenience and straight dye fronts
- Easy to use — no clamps, no loose parts, bottom drain port
- Utilizes internal pump for even temperature distribution
- Compatible with a recirculating chiller for active cooling
Certifications
UL61010-1, CSAC22.2 No. 61010-1, EN61010-1:2001, CE Marked
Specifications
EN61010-1, UL61010-1, CSA C22.2, CE | |
113°F | |
15°C to 30°C | |
9.84 in. | |
25 cm | |
7.87 in. | |
7.87 in. | |
29.98 lb. | |
600 V | |
1000 mA |
Up to 80% | |
45°C | |
16.92 in. | |
43 cm | |
16.92 in. | |
43 cm | |
20 cm | |
13.6 kg | |
150 w |