missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SE600X Chroma Deluxe Dual Cooled Vertical Electrophoresis Unit

Run up to four gels at a time under identical conditions

Brand:  Hoefer™ SE600X

Product Code. 12855836

  • £1635.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket

Includes: Lower buffer chamber (tank), safety lid with leads, upper buffer chamber with cathode, heat-exchanger with anode, 3 sets of glass plates (18 x 16cm), four 16cm clamp assemblies, 6 cams, dual gel casting stand with leveling base and level, buffer dam, Spacer-Mate alignment template, Wonder Wedge plate separation tool, Gel Seal.
This item is not returnable. View return policy

Description

Description

  • Rounded contours allow for easy handling, lifting, introducing sample, and disassembling
  • Produces straight lanes and sharp, well-defined bands
  • Runs as many as four gels (maximum 112 samples) at one time under identical conditions (with accessory Club Sandwich Kit)
  • Accepts gels 8 cm or 16 cm long
  • Accommodates denaturing and native polyacrylamide gels, and the second dimension of 2-D electrophoresis
  • A wide variety of accessories enables you to tailor gel configurations to your needs
  • Runs gels at temperatures from 1 to 45°C
  • Storage temperature: +15 °C to +30°C
  • Does not come with combs and spacers; order two SE511 combs and two sets of 16 cm long spacers
Specifications

Specifications

EN61010-1, UL61010-1, CSA C22.2 1010.1, CE
Lower Buffer chamber, upper Buffer chamber, safety lid with high voltage Leads, heat exchanger, Glass plates, clamp assemblies, cams, slotted gaskets for upper Buffer chamber, dual gel casting stand with leveling base, bubble level, laminated gaskets for
45°C
12.59 in.
32 cm
11.41 in.
29 cm
14 cm
50 w
Up to 80%
113°F
15°C to 30°C
7.08 in.
18 cm
3.14 to 6.29 in.
5.51 in.
1000 V
500 mA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers