missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SG Series Gradient Makers

Ideal for generating gradients of polyacrylamide, sucrose, and cesium chloride

Brand:  Hoefer™ SG100

 View more versions of this product

Product Code. 11564424

  • £515.00 / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket

Includes:
  • Models SG 15, SG 30, SG 50, and SG 100 include gradient maker with PTFE valves, Luer outlet fitting, support rod, adapter barbed fitting, 22-gauge needle.
  • Model SG 500 includes gradient maker with push-pull valve, four wrap-and-seal hose clamps.
This item is not returnable. View return policy

Description

Description

Models SG 15, SG 30, SG 50, and SG 100

  • Milled of heavy acrylic plastic with high-quality, leak-free PTFE-resin valves
  • Offer a flat base for stability on magnetic stirrers; support rod can be attached to ring stand for additional stability
  • Accept Luer or 4 mm ID tubing; with included adapters can also accept 2 mm ID or 22-gauge tubing
  • SG 100 is ideal for the Hoefer Mighty Small™ Gel Casters SE 275 (for four gels) or SE 215 (for 10 gels)
Model SG 500
  • Made of two cylindrical acrylic chambers joined and mounted on a flat acrylic base
  • Uses a simple push-pull valve to open and close the passage between the reservoir and mixing chambers
  • Suitable for forming polyacrylamide gradients or buffer gradients used with medium to small chromatography columns
  • Ideal for use with the Hoefer SE 615 or SE 915 Multiple Gel Casters
Specifications

Specifications

Mighty Small™ Gel Caster SE 275 (for 4 Gels) or SE 215 (for 10 Gels)
Gradient maker w/PTFE Valves , Luer outlet fitting, support rod, adapter barbed fitting and 22G needle
100 mL
Gradient Maker
3.38 oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hoefer™ SG Series Gradient Makers > Model SG 100; 100mL total volume

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.