Learn More
Hoefer™ SUB15 Standard Submarine Combs
Accessories for SUB15 Standard Submarine
£67.39 - £70.52
Specifications
For Use With (Equipment) | SUB15 Standard Submarine |
---|---|
Type | Comb |
Depth (English) Pan | 0.19 in. |
Depth (Metric) Pan | 0.5 cm |
Product Code | Brand | Thickness (English) | Thickness (Metric) | No. of Wells | Width (Metric) Well | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (English) | Thickness (Metric) | No. of Wells | Width (Metric) Well | Price | Quantity & Availability | ||
12695760
|
Hoefer™
SUB15-C1.5-10 |
0.05 in. | 0.15 cm | 10 | 1.28 cm |
£67.39
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12605770
|
Hoefer™
SUB15-C1.5-16MC |
0.05 in. | 0.15 cm | 16 | 0.64 cm |
£67.39
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12615770
|
Hoefer™
SUB15-C1.5-20 |
0.05 in. | 0.15 cm | 20 | 0.59 cm |
£67.39
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12625770
|
Hoefer™
SUB15-C1.5-30MC |
0.05 in. | 0.15 cm | 30 | 0.25 cm |
£67.39
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12635756
|
Hoefer™
SUB15-C1.5-25 |
0.05 in. | 0.15 cm | 25 | 0.35 cm |
£67.39
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12655756
|
Hoefer™
SUB15-C1-16MC |
0.03 in. | 0.1 cm | 16 | 0.64 cm |
£67.39
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12635770
|
Hoefer™
SUB15-C2-10 |
0.07 in. | 0.2 cm | 16 | 1.28 cm |
£70.52
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12645756
|
Hoefer™
SUB15-C1-10 |
0.03 in. | 0.1 cm | 10 | 1.28 cm |
£70.52
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12655770
|
Hoefer™
SUB15-C2-20 |
0.07 in. | 0.2 cm | 20 | 0.59 cm |
£67.39
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12675756
|
Hoefer™
SUB15-C1-25 |
0.03 in. | 0.1 cm | 25 | 0.35 cm |
£70.52
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12695756
|
Hoefer™
SUB15-C2-25 |
0.07 in. | 0.2 cm | 25 | 0.35 cm |
£70.52
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12665756
|
Hoefer™
SUB15-C1-20 |
0.03 in. | 0.1 cm | 20 | 0.59 cm |
£70.52
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12645770
|
Hoefer™
SUB15-C2-16MC |
0.07 in. | 0.2 cm | 20 | 0.64 cm |
£70.52
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12685756
|
Hoefer™
SUB15-C1-30MC |
0.03 in. | 0.1 cm | 30 | 0.25 cm |
£67.39
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
12665770
|
Hoefer™
SUB15-C2-30MC |
0.07 in. | 0.2 cm | 30 | 0.25 cm |
£67.39
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||
Description
- Ranges from 10, 16, 20, 25 and 30 Wells
- 1, 1.5 and 2 mm Thickness
- 2.5 to 12.8 mm Well Width
- Min. 12, max. 128 μL Sample Volume
Specifications
SUB15 Standard Submarine | |
Comb | |
0.19 in. | |
0.5 cm |