Learn More
Hoefer™ SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarine Combs
Accessories for SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarines
Brand:  Hoefer™ SUB20-C1-40MC
 View more versions of this product
Product Code. 12645776
  • £67.39 / Each
Explore more special offers
Description
Description
  • Ranges from 16, 20, 28 and 40 Wells
  • 1, 1.5 and 2 mm Thickness
  • 3, 5, 7 and 8.5 mm Well Width
  • Min. 13, max. 75 μL Sample Volume
Specifications
Specifications
SUB20 Maxi Standard and SUB20C Maxi Cooled Submarine
40
0.1 cm
0.11 in.
0.19 in.
Comb
0.03 in.
13 μL
0.3 cm
0.5 cm