Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Electrode Panels
For use with TE 42 and TE 62 Transfer Tank
Brand:  Hoefer™ TE43GY
 View more versions of this product
Product Code. 12099857
  • £410.00 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Comes in gray or black electrode panels
  • Type: Gray Electrode Panel
  • Includes Banana plug and platinum wire
Specifications
Specifications
Gray
TE 42, TE 62 Transfer Tank
Gray Electrode Panel
Banana plug and Platinum wire