missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ TE70X and TE77X Semidry Blotting Systems
Perform uniform transfers of proteins and nucleic acids from polyacrylamide and agarose gels in less than one hour
£1277.23 - £1723.87
Specifications
Certifications/Compliance | EN61010-1, UL3101-1, CSA C22.2 1010.1, CE |
---|---|
For Use With (Equipment) | Electrophoresis Blotting System |
Type | Semi Dry Transfer Unit |
Humidity Range | Up to 80% |
Temperature (English) Maximum | 113°F |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11905348
|
Hoefer™
TE70XP |
Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (14 x 16cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™ |
£1692.52
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
11915348
|
Hoefer™
TE77XP |
Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™ |
£1723.87
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
11995338
|
Hoefer™
TE77X |
Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™ masks (16.5 x |
£1277.23
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
Description
- Transfer up to two polyacrylamide gels in a stack
- Use minimal buffer
- Do not generate the excessive heat that can distort bands or denature protein
- Utilize durable iridium oxide and stainless-steel electrodes to assure uniform, contamination-free transfers
- Offer automatic transfer-stopping feature to prevent overheating when buffer is depleted
- Available with built-in power supply to save space and simplify transfer
Certifications
CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1
Specifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE | |
Semi Dry Transfer Unit | |
113°F | |
18.89 x 15 x 4 in. (48 x 38 x 10 cm) | |
48 cm | |
38 cm | |
10 cm | |
15 w |
Electrophoresis Blotting System | |
Up to 80% | |
45°C | |
18.89 in. | |
15 in. | |
4 in. | |
30 V | |
500 mA |