missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Holmium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Holmium(III) acetate hydrate is used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline Aggregates |
Quantity | 50 g |
Solubility Information | Soluble in water. |
Formula Weight | 342.07 (Anhydrous) |
Percent Purity | 99.9% |
Grade | Reacton™ |
Assay | (REO) |
Chemical Name or Material | Holmium(III) acetate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'