missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Holmium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Holmium(III) fluoride is used as an intermediate in the manufacture of glass, metal halide lamp and dopant to garnet laser. It is employed in metallurgy, chemical and physical vapor deposition and in some optical coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 1143°C |
Physical Form | Crystalline Lump |
Quantity | 5 g |
UN Number | UN3288 |
Solubility Information | Insoluble in water. |
Formula Weight | 221.93 |
Percent Purity | 99.99% |
Grade | Reacton™ |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'