missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh, Thermo Scientific Chemicals

Product Code. 10247023
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 12055-62-8
Molecular Formula Ho2O3
Molecular Weight (g/mol) 377.86
MDL Number MFCD00011053
InChI Key OWCYYNSBGXMRQN-UHFFFAOYSA-N
IUPAC Name diholmium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ho+3].[Ho+3]

Specifications

Boiling Point 3900.0°C
Melting Point 2330.0°C
Color Beige to Orange-Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.85% min. (Ho2O3) TREO base
Linear Formula HO2O3
Merck Index 15, 4765
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in inorganic acids
Formula Weight 377.85
Percent Purity 99.9%
Packaging Glass bottle
Chemical Name or Material Holmium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468819 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number197140100,197140500
Lot NumberA0431931 Certificate TypeCertificate of Analysis Date01/01/2024 Catalog Number197140100,197140500
Lot NumberA0458378 Certificate TypeCertificate of Analysis Date01/12/2023 Catalog Number197140100,197140500
Lot NumberA0449009 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number197140100,197140500
Lot NumberA0400744 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number197140100,197140500
Product Identifier
  • Holmium(III) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.