Learn More
Holmium(III) oxide, REacton™, 99.995% (REO)
Description
Holmium(III) oxide iacts as colorants which is used for cubic zirconia and glass, as a calibration standard for optical spectrophotometers, as a specialty catalyst, phosphor and a laser material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2360°C |
| Color | Beige |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.995% (REO) |
| Solubility Information | Insoluble in water. Soluble in perchloric acids. |
| Formula Weight | 377.86 |
| Odor | Odorless |
| Appearance | Beige powder or lump |
| Chemical Name or Material | Holmium(III) oxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.