missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homovanillic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 141°C to 145°C |
Color | White to Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Linear Formula | HOC6H3(OCH3)CH2CO2H |
Quantity | 1 g |
Beilstein | 10, 409 |
Merck Index | 15, 4779 |
Solubility Information | Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,ether,soluble in benzene,insoluble in cyclohexane |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'