Learn More
Sperian™ Honeywell™ 2000 Comfort Series Respirators
With an ergonomic design to deliver outstanding levels of comfort and adjustment. Sperian™ Honeywell™ 2000 Comfort Series Respirators have a cut-out shape to provide excellent compatibility with goggles.
Brand:  Sperian™ 1031590
 View more versions of this product
Product Code. 15341141
  • £42.45 / Pack of 25
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Folds flat packed in individually sealed bags for transportation and storage
  • Dolomite tested, demonstrating the respirators' resistance to clogging over time
  • With high efficiency exhalation valve for protection
  • Double layer face seal provides comfort and protection
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN 149 : 2001 A1: 2009
FFP1
Medium
Agriculture, Chemical, Construction, Fire Protection, Food Services, Forestry, Government, Manufacturing, Mining, Municipal Services, Oil and Gas, Pharmaceutical, Steel and Metals, Utilities
2110 FFP1
Flat Fold
Single-use
Respirator Protection
Yes
Yes
25/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sperian™ Honeywell™ 2000 Comfort Series Respirators > Model: 2110 FFP1