Learn More
Sperian™ Honeywell™ Santa Cruz Safety Glasses Product Code.: 12602743
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
Sperian™ Honeywell™ Santa Cruz Safety Glasses
Product Code. 12602743
Blue
Click to view available options
Frame Color:
Blue
Red
Product Code. 12602743
Brand: Sperian™ 1002676
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
Add to basket
A comfortable, stylish, lightweight spectacle with a one-piece wrap-around lens. Sperian™ Honeywell™ Santa Cruz Safety Glasses provide unobstructed wide-angle vision.
  • Adjustable positive-lock temples
  • Polycarbonate lenses
  • Provides 99.9% UV protection
  • Available in blue or red 
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001 / 2000, EC EC0165
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Eye and Face Protection Spectacles
For Use With (Application) UV protection glasses
Frame Color Blue
No. per Pack 1 Ea.
Lens Material Polycarbonate
Frame Material Nylon
Product Content Correction
Product Title
Sperian™ Honeywell™ Santa Cruz Safety Glasses >