Learn More
Sperian™ Honeywell™ Matrix Uncorded Earplugs
missing translation for 'orderingAttributeHoverText'
Color:
Blue
Orange
Description
- Available in either blue or orange
- Uniform attenuation profile blocks loud noise while voice and other higher frequencies can be heard naturally
- Smaller diameter eliminated that "plugged up" feeling in the ear canal
- Smooth outer skin is a great match for the sensitive skin inside the ear canal
Specifications
Specifications
Color | Blue |
Size | Universal |
Quantity | 200/Pk. |
Plug Shape | Cylinder |
Product Type | Earplug |
Disposable | Single-use |